The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172557.2 | NP_766145.1 | 14 - 15 | 1782 | 65 |
GenBank mRNA | AJ250024.1 | - | 13 - 14 | 1458 | 65 |
AK046633.1 | - | 14 - 15 | 1783 | 65 |