Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tada2a
Entrez Gene ID
217031
Gene Name
transcriptional adaptor 2A
Gene Aliases
AV319371, D030022J10Rik, Tada2l
Chromosome Location
Chr.11: 84078920 - 84129642 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172562.3 NP_766150.1 3 - 4 247 143
GenBank mRNA AK142262.1 - 3 - 4 279 143
AK157938.1 - 3 - 4 240 143
BC038821.1 - 3 - 4 250 143

View Full Product Details