Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Trim47
Entrez Gene ID
217333
Gene Name
tripartite motif-containing 47
Gene Aliases
2210023F24Rik
Chromosome Location
Chr.11: 116105750 - 116110235 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205081.1 NP_001192010.1 2 - 3 802 83
NM_172570.5 NP_766158.3 2 - 3 802 83
GenBank mRNA AK031572.1 - 2 - 3 777 83
AK075838.1 - 2 - 3 231 83
BC043714.1 - 1 - 2 114 83
BC064728.1 - 2 - 3 196 83
BC080731.1 - 2 - 3 198 83
BC100317.1 - 2 - 3 786 83
BC132148.1 - 2 - 3 583 83
BC137953.1 - 2 - 3 583 83
BC145059.1 - 2 - 3 581 83

View Full Product Details