Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mis18bp1
Entrez Gene ID
217653
Gene Name
MIS18 binding protein 1
Gene Aliases
6720403H11, C79407, Knl2, mKIAA1903
Chromosome Location
Chr.12: 65132734 - 65172580 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172578.2 NP_766166.2 10 - 11 2819 127
GenBank mRNA AK032698.1 - 8 - 9 1851 127
AK035117.1 - 10 - 11 2816 127
AK220223.1 - 9 - 10 2085 127
BC021832.1 - 5 - 6 1332 127
BC052175.1 - 10 - 11 2819 127

View Full Product Details