The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172582.3 | NP_766170.2 | 2 - 3 | 407 | 58 |
GenBank mRNA | AK077863.1 | - | 2 - 3 | 331 | 58 |
AK131889.1 | - | 2 - 3 | 369 | 58 | |
AK167380.1 | - | 2 - 3 | 335 | 58 | |
AK170311.1 | - | 2 - 3 | 335 | 58 | |
BC024135.1 | - | 2 - 3 | 308 | 58 | |
BC051055.1 | - | 2 - 3 | 323 | 58 | |
BC088736.1 | - | 2 - 3 | 379 | 58 | |
BC106183.1 | - | 2 - 3 | 344 | 58 | |
BC132168.1 | - | 2 - 3 | 407 | 58 | |
BC132170.1 | - | 2 - 3 | 407 | 58 |