Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem63c
Entrez Gene ID
217733
Gene Name
transmembrane protein 63c
Gene Aliases
4932420N09, 9330187M14Rik
Chromosome Location
Chr.12: 87021344 - 87091824 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172583.2 NP_766171.1 23 - 24 2445 99
GenBank mRNA AK030062.1 - 23 - 24 2437 99
AK034410.1 - 22 - 23 2368 99
AK043536.1 - 22 - 23 2367 99
BC056936.1 - 23 - 24 2445 99
BC057088.1 - 23 - 24 2445 99

View Full Product Details