Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arl15
Entrez Gene ID
218639
Gene Name
ADP-ribosylation factor-like 15
Gene Aliases
A430036I03, Arfrp2, C230032K13Rik
Chromosome Location
Chr.13: 113794506 - 114157462 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172595.4 NP_766183.1 2 - 3 336 59
GenBank mRNA AK039965.1 - 2 - 3 300 59
AK048736.1 - 2 - 3 219 59
AK050979.1 - 2 - 3 251 59
AK052164.1 - 2 - 3 285 59
AK053953.1 - 3 - 4 403 59
BC120664.1 - 2 - 3 222 59
BC120666.1 - 2 - 3 222 59
CJ063085.1 - 2 - 3 336 59
CJ066332.1 - 2 - 3 320 59

View Full Product Details