The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172618.2 | NP_766206.1 | 11 - 12 | 2038 | 63 |
GenBank mRNA | AK052664.1 | - | 11 - 12 | 1922 | 63 |
BC057897.1 | - | 11 - 12 | 1916 | 63 |