Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Glis3
Entrez Gene ID
226075
Gene Name
GLIS family zinc finger 3
Gene Aliases
4833409N03Rik, E330013K21Rik
Chromosome Location
Chr.19: 28262179 - 28680077 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK054020.1 - 2 - 3 972 58
DQ871027.1 - 2 - 3 972 58

View Full Product Details