Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kctd3
Entrez Gene ID
226823
Gene Name
potassium channel tetramerisation domain containing 3
Gene Aliases
4930438A20Rik, 9330185B06, E330032J19Rik, NY-REN-45
Chromosome Location
Chr.1: 188971095 - 189007868 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172650.2 NP_766238.1 10 - 11 1168 100
GenBank mRNA AK033491.1 - 10 - 11 1169 100
AK034380.1 - 10 - 11 1169 100
AK049362.1 - 10 - 11 1166 100
AK087865.1 - 10 - 11 1169 100
AK139700.1 - 10 - 11 1169 100
BC061458.1 - 10 - 11 1156 100
BC079581.1 - 10 - 11 1155 100

View Full Product Details