Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc2a6
Entrez Gene ID
227659
Gene Name
solute carrier family 2 (facilitated glucose transporter), member 6
Gene Aliases
A330096C23, F630103L12Rik, Glut6, Glut9
Chromosome Location
Chr.2: 27021363 - 27028001 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172659.2 NP_766247.2 7 - 8 1123 59
GenBank mRNA AK079650.1 - 7 - 8 1067 59
AK089246.1 - 7 - 8 1115 59
AK137441.1 - 6 - 7 1338 59
AK150104.1 - 7 - 8 1076 59
AK163445.1 - 7 - 8 1081 59
AK166312.1 - 7 - 8 1081 59
BC025498.1 - 4 - 5 1707 59
BC145480.1 - 7 - 8 1049 59

View Full Product Details