Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Acad9
Entrez Gene ID
229211
Gene Name
acyl-Coenzyme A dehydrogenase family, member 9
Gene Aliases
2600017P15Rik, 4732402K02, C630012L17Rik, NPD002
Chromosome Location
Chr.3: 36065984 - 36092857 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172678.3 NP_766266.3 11 - 12 1268 63
GenBank mRNA AK028571.1 - 10 - 11 1982 63
AK031820.1 - 11 - 12 1200 63
AK075984.1 - 11 - 12 1187 63
AK145423.1 - 11 - 12 1176 63
AK205472.1 - 1 - 2 86 63
AK212715.1 - 2 - 3 159 63
BC004737.1 - 5 - 6 514 63
BC031137.1 - 11 - 12 1204 63
BC032213.1 - 11 - 12 1209 63
BC033277.1 - 11 - 12 1189 63

View Full Product Details