The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172713.2 | NP_766301.1 | 16 - 17 | 1500 | 60 |
GenBank mRNA | AK029881.1 | - | 16 - 17 | 1502 | 60 |
AK160646.1 | - | 16 - 17 | 1368 | 60 | |
BC042708.1 | - | 16 - 17 | 1450 | 60 |