Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Brat1
Entrez Gene ID
231841
Gene Name
BRCA1-associated ATM activator 1
Gene Aliases
AA881470, Baat1
Chromosome Location
Chr.5: 140704786 - 140719379 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276287.1 NP_001263216.1 8 - 9 1314 98
NM_181066.3 NP_851411.1 8 - 9 1216 98
NR_074085.1 7 - 8 1124 98
GenBank mRNA AK043212.1 - 8 - 9 1203 98
AK049321.1 - 7 - 8 1113 98
AK084987.1 - 8 - 9 1205 98
AK085091.1 - 8 - 9 1205 98
AK142387.1 - 5 - 6 2472 98
AK154448.1 - 8 - 9 98
AK170269.1 - 8 - 9 1210 98
BC068301.1 - 8 - 9 1280 98

View Full Product Details