Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fgd5
Entrez Gene ID
232237
Gene Name
FYVE, RhoGEF and PH domain containing 5
Gene Aliases
C330025N11Rik, ZFYVE23, mFLJ00274
Chromosome Location
Chr.6: 91978821 - 92076005 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172731.2 NP_766319.2 14 - 15 3506 110
GenBank mRNA AK049341.1 - 1 - 2 180 110
AK220153.1 - 15 - 16 3980 110
BC042732.1 - 10 - 11 923 110
BC060664.1 - 14 - 15 2969 110
BC145502.1 - 15 - 16 3323 110
BC150786.1 - 14 - 15 3455 110

View Full Product Details