Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Fbxl19
- Entrez Gene ID
-
233902
- Gene Name
- F-box and leucine-rich repeat protein 19
- Gene Aliases
-
6030430O11, BC059812, Fbl19
- Chromosome Location
-
Chr.7: 127744027 - 127769483 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK031431.1 |
- |
4 - 5 |
571 |
78 |
View Full Product Details