The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252623.1 | NP_001239552.1 | 6 - 7 | 1853 | 61 |
NM_172753.5 | NP_766341.4 | 5 - 6 | 1549 | 61 | |
GenBank mRNA | AK028688.1 | - | 5 - 6 | 1472 | 61 |
AK033522.1 | - | 3 - 4 | 1203 | 61 | |
AK049770.1 | - | 6 - 7 | 2147 | 61 | |
AK080687.1 | - | 5 - 6 | 1518 | 61 | |
AK133567.1 | - | 3 - 4 | 1202 | 61 | |
AK144089.1 | - | 2 - 3 | 1110 | 61 | |
BC057630.1 | - | 7 - 8 | 1821 | 61 | |
BC089162.1 | - | 5 - 6 | 1546 | 61 |