Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nfrkb
Entrez Gene ID
235134
Gene Name
nuclear factor related to kappa B binding protein
Gene Aliases
A530090G11Rik
Chromosome Location
Chr.9: 31386161 - 31421333 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172766.3 NP_766354.2 17 - 18 1966 106
GenBank mRNA AK041206.1 - 17 - 18 2204 106
AK049804.1 - 3 - 4 341 106
BC033595.1 - 17 - 18 1956 106
BC059861.1 - 17 - 18 2013 106
BC059900.1 - 8 - 9 1613 106
BC060076.1 - 17 - 18 2195 106

View Full Product Details