The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172780.3 | NP_766368.2 | 9 - 10 | 1267 | 117 |
GenBank mRNA | AK032326.1 | - | 9 - 10 | 1248 | 117 |
AK122232.1 | - | 9 - 10 | 1238 | 117 | |
BC130221.1 | - | 9 - 10 | 1252 | 117 |