Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhl4
Entrez Gene ID
237010
Gene Name
kelch-like 4
Gene Aliases
B130014A11, C130018J01Rik
Chromosome Location
Chr.X: 114474333 - 114561129 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290477.1 NP_001277406.1 8 - 9 1776 69
NM_001290478.1 NP_001277407.1 8 - 9 1695 69
NM_172781.2 NP_766369.2 8 - 9 1794 69
GenBank mRNA AK044930.1 - 8 - 9 1426 69
AK081448.1 - 8 - 9 1778 69
AK081937.1 - 8 - 9 1607 69
AK085099.1 - 8 - 9 1777 69
AK161786.1 - 8 - 9 1670 69
BC046395.1 - 2 - 3 354 69
BC057137.1 - 8 - 9 1525 69
BC141251.1 - 8 - 9 1519 69

View Full Product Details