The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168521.1 | NP_001161993.1 | 7 - 8 | 2337 | 66 |
NM_172795.3 | NP_766383.2 | 7 - 8 | 2217 | 66 | |
GenBank mRNA | AK044113.1 | - | 2 - 3 | 554 | 66 |
AK172964.1 | - | 6 - 7 | 2480 | 66 | |
AY444167.1 | - | 7 - 8 | 2041 | 66 | |
BC052098.1 | - | 7 - 8 | 1863 | 66 | |
BC058534.1 | - | 7 - 8 | 2164 | 66 | |
BC080850.1 | - | 7 - 8 | 2217 | 66 | |
BU704378.1 | - | 3 - 4 | 588 | 66 |