The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253759.1 | NP_001240688.1 | 6 - 7 | 816 | 57 |
NM_172813.3 | NP_766401.1 | 6 - 7 | 816 | 57 | |
GenBank mRNA | AK051821.1 | - | 6 - 7 | 764 | 57 |
AK082388.1 | - | 6 - 7 | 817 | 57 | |
BC052062.1 | - | 3 - 4 | 327 | 57 |