Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lipk
Entrez Gene ID
240633
Gene Name
lipase, family member K
Gene Aliases
9930115F20, BC055815, Lipl2
Chromosome Location
Chr.19: 34008254 - 34047903 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172837.4 NP_766425.1 2 - 3 235 89
GenBank mRNA AK037106.1 - 2 - 3 200 89
AK162706.1 - 2 - 3 157 89
BY150606.1 - 2 - 3 235 89

View Full Product Details