The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172838.3 | NP_766426.1 | 3 - 4 | 401 | 102 |
GenBank mRNA | AK028284.1 | - | 3 - 4 | 399 | 102 |
AK029076.1 | - | 3 - 4 | 401 | 102 | |
BC111902.1 | - | 2 - 3 | 354 | 102 | |
BC113803.1 | - | 2 - 3 | 354 | 102 |