The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172843.4 | NP_766431.3 | 6 - 7 | 1696 | 81 |
GenBank mRNA | AK038024.1 | - | 6 - 7 | 1958 | 81 |
AK080959.1 | - | 4 - 5 | 975 | 81 | |
BC066790.1 | - | 4 - 5 | 902 | 81 | |
CK635751.1 | - | 2 - 3 | 537 | 81 |