Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Pak7
- Entrez Gene ID
-
241656
- Gene Name
- p21 protein (Cdc42/Rac)-activated kinase 7
- Gene Aliases
-
2900083L08Rik, 6430627N20, Pak5
- Chromosome Location
-
Chr.2: 136081088 - 136387967 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK079080.1 |
- |
1 - 2 |
285 |
58 |
View Full Product Details