Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Svs1
Entrez Gene ID
243377
Gene Name
seminal vesicle secretory protein 1
Gene Aliases
9530003A11
Chromosome Location
Chr.6: 48986861 - 48991725 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172888.3 NP_766476.2 4 - 5 2630 83
GenBank mRNA AK035222.1 - 4 - 5 2631 83
AY115568.1 - 4 - 5 2587 83
AY283179.1 - 4 - 5 2587 83
BC125635.1 - 4 - 5 2501 83

View Full Product Details