Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Styk1
Entrez Gene ID
243659
Gene Name
serine/threonine/tyrosine kinase 1
Gene Aliases
9130025L13, AI326477, Nok
Chromosome Location
Chr.6: 131299133 - 131317009 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172891.2 NP_766479.1 8 - 9 1932 103
GenBank mRNA AK033606.1 - 8 - 9 1932 103
AK035247.1 - 8 - 9 1093 103
AY563053.1 - 8 - 9 1087 103
BC150724.1 - 8 - 9 1140 103

View Full Product Details