The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172911.3 | NP_766499.2 | 1 - 2 | 163 | 143 |
GenBank mRNA | AK036672.1 | - | 1 - 2 | 131 | 143 |
AK052494.1 | - | 1 - 2 | 131 | 143 | |
AK089749.1 | - | 1 - 2 | 131 | 143 |