Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pcnxl2
Entrez Gene ID
270109
Gene Name
pecanex-like 2 (Drosophila)
Gene Aliases
A830048P05, AU067631, E330039K12Rik, mKIAA0435
Chromosome Location
Chr.8: 125751504 - 125898317 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175561.4 NP_780770.2 14 - 15 3156 73
GenBank mRNA AK043903.1 - 12 - 13 2517 73
AK220342.1 - 6 - 7 718 73
BC132334.1 - 10 - 11 2379 73

View Full Product Details