The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170978.1 | NP_001164449.1 | 4 - 5 | 391 | 118 |
NM_172961.3 | NP_766549.2 | 4 - 5 | 391 | 118 | |
GenBank mRNA | AK036128.1 | - | 4 - 5 | 336 | 118 |
AK132282.1 | - | 4 - 5 | 118 | ||
AK160802.1 | - | 4 - 5 | 339 | 118 | |
AK165348.1 | - | 4 - 5 | 341 | 118 | |
BC058079.1 | - | 4 - 5 | 331 | 118 | |
BC058521.1 | - | 4 - 5 | 331 | 118 |