The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166033.1 | NP_001159505.1 | 3 - 4 | 2409 | 101 |
NM_172977.3 | NP_766565.2 | 2 - 3 | 2303 | 101 | |
GenBank mRNA | AK030351.1 | - | 2 - 3 | 2230 | 101 |
AK047698.1 | - | 1 - 2 | 1413 | 101 | |
AK141269.1 | - | 2 - 3 | 2285 | 101 | |
AK146401.1 | - | 3 - 4 | 2365 | 101 | |
AK146403.1 | - | 3 - 4 | 2365 | 101 | |
AK150650.1 | - | 2 - 3 | 2285 | 101 | |
AK151179.1 | - | 2 - 3 | 2285 | 101 | |
AK153102.1 | - | 2 - 3 | 2285 | 101 | |
BC095998.1 | - | 2 - 3 | 2242 | 101 | |
BC117919.1 | - | 2 - 3 | 2215 | 101 | |
BC117920.1 | - | 2 - 3 | 2213 | 101 |