Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rnft2
Entrez Gene ID
269695
Gene Name
ring finger protein, transmembrane 2
Gene Aliases
AW049082, B830028P19Rik, Tmem118
Chromosome Location
Chr.5: 118190734 - 118245111 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172998.3 NP_766586.2 1 - 2 30 62
GenBank mRNA AK046850.1 - 1 - 2 31 62

View Full Product Details