The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109902.1 | NP_001103372.1 | 11 - 11 | 2615 | 99 |
NM_172998.3 | NP_766586.2 | 12 - 12 | 2650 | 99 | |
GenBank mRNA | AK046850.1 | - | 12 - 12 | 2652 | 99 |
AK148930.1 | - | 11 - 11 | 2594 | 99 | |
BC057302.1 | - | 11 - 11 | 2577 | 99 | |
BU611972.1 | - | 1 - 1 | 430 | 99 |