The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173070.1 | NP_775093.1 | 1 - 1 | 199 | 64 |
GenBank mRNA | AY158996.1 | - | 1 - 1 | 199 | 64 |
BC107019.1 | - | 1 - 1 | 199 | 64 |