The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173384.2 | NP_775560.1 | 1 - 2 | 1061 | 117 |
GenBank mRNA | AY005801.1 | - | 1 - 2 | 1061 | 117 |