The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199283.1 | NP_001186212.1 | 8 - 9 | 1277 | 57 |
NM_001199284.1 | NP_001186213.1 | 8 - 9 | 1059 | 57 | |
NM_173388.2 | NP_775564.1 | 8 - 9 | 1022 | 57 | |
GenBank mRNA | AB120363.1 | - | 8 - 9 | 1055 | 57 |
AK143773.1 | - | 8 - 9 | 1120 | 57 | |
AK159144.1 | - | 8 - 9 | 1245 | 57 | |
AK171104.1 | - | 8 - 9 | 1279 | 57 | |
AK172685.1 | - | 8 - 9 | 1049 | 57 | |
BC042513.1 | - | 8 - 9 | 1018 | 57 |