The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173391.3 | NP_775567.2 | 4 - 5 | 667 | 82 |
GenBank mRNA | AY090565.1 | - | 4 - 5 | 667 | 82 |
BC120514.1 | - | 4 - 5 | 658 | 82 | |
BC125430.1 | - | 4 - 5 | 658 | 82 |