Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tph2
Entrez Gene ID
216343
Gene Name
tryptophan hydroxylase 2
Gene Aliases
AU043594, Ntph
Chromosome Location
Chr.10: 115078641 - 115185022 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173391.3 NP_775567.2 5 - 6 737 85
GenBank mRNA AY090565.1 - 5 - 6 737 85
BC120514.1 - 5 - 6 728 85
BC125430.1 - 5 - 6 728 85

View Full Product Details