Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nyx
Entrez Gene ID
236690
Gene Name
nyctalopin
Gene Aliases
CLNP, CSNB1, CSNB4, nob
Chromosome Location
Chr.X: 13461095 - 13489314 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173415.4 NP_775591.1 1 - 2 153 109
GenBank mRNA BC035244.1 - 1 - 2 153 109
BC048381.1 - 1 - 2 152 109

View Full Product Details