The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173439.2 | NP_775615.2 | 1 - 2 | 530 | 57 |
GenBank mRNA | BC026799.1 | - | 1 - 2 | 602 | 57 |
BC088738.1 | - | 1 - 2 | 530 | 57 | |
BC098203.1 | - | 1 - 2 | 607 | 57 |