The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173450.3 | NP_775626.1 | 1 - 2 | 678 | 76 |
GenBank mRNA | AK014804.1 | - | 1 - 2 | 638 | 76 |
AK035752.1 | - | 1 - 2 | 617 | 76 | |
AK035800.1 | - | 1 - 2 | 617 | 76 | |
AK043340.1 | - | 1 - 2 | 646 | 76 | |
AK044767.1 | - | 1 - 2 | 679 | 76 | |
BC117827.1 | - | 1 - 2 | 679 | 76 | |
BC117828.1 | - | 1 - 2 | 679 | 76 |