The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_174988.3 | NP_778153.2 | 4 - 5 | 1154 | 86 |
GenBank mRNA | AB019618.1 | - | 4 - 5 | 1144 | 86 |
AK140248.1 | - | 4 - 5 | 1272 | 86 | |
BC112407.1 | - | 3 - 4 | 849 | 86 |