The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_174988.3 | NP_778153.2 | 11 - 12 | 2400 | 67 |
GenBank mRNA | AB019618.1 | - | 11 - 12 | 2390 | 67 |
AK140248.1 | - | 11 - 12 | 2518 | 67 | |
BC112407.1 | - | 10 - 11 | 2033 | 67 |