The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175090.4 | NP_780299.2 | 3 - 4 | 401 | 110 |
GenBank mRNA | AK046835.1 | - | 3 - 4 | 354 | 110 |
AK151337.1 | - | 3 - 4 | 383 | 110 | |
AK170285.1 | - | 3 - 4 | 354 | 110 | |
BC034674.1 | - | 3 - 4 | 339 | 110 | |
BC046470.1 | - | 1 - 2 | 75 | 110 | |
BC058227.1 | - | 3 - 4 | 341 | 110 | |
BC065147.1 | - | 2 - 3 | 249 | 110 |