Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdca2
Entrez Gene ID
108912
Gene Name
cell division cycle associated 2
Gene Aliases
2610311M19Rik, AI586158
Chromosome Location
Chr.14: 67673180 - 67715841 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110162.1 NP_001103632.1 4 - 5 781 92
NM_175384.4 NP_780593.3 4 - 5 484 92
GenBank mRNA AK031083.1 - 4 - 5 547 92
AK160223.1 - 4 - 5 543 92
AK166952.1 - 4 - 5 550 92
BC037136.1 - 4 - 5 581 92
BC067009.1 - 4 - 5 512 92
BC075682.1 - 4 - 5 484 92

View Full Product Details