Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Camk2a
Entrez Gene ID
12322
Gene Name
calcium/calmodulin-dependent protein kinase II alpha
Gene Aliases
CaMKII, R74975, mKIAA0968
Chromosome Location
Chr.18: 60925580 - 60988152 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177407.4 NP_803126.1 3 - 4 363 65
GenBank mRNA AK083245.1 - 3 - 4 364 65
AK122410.2 - 3 - 4 359 65
BC031745.1 - 3 - 4 344 65
X14836.1 - 3 - 4 347 65

View Full Product Details