Product Details

Assay Reference Genome
Location

Chr.9:59564861 on build GRCm38
Cytoband
9B
Species
Mus musculus
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
15211
Gene Symbol
Hexa
Gene Name
hexosaminidase A
Gene Aliases
Hex-1
Location
Chr.9:59539545-59565109 on build GRCm38
Assay Gene Location
Within Exon 14
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
Hexa NM_010421.4 14 1621 NP_034551.2
AK075895.1 14 1597 BAC36036.1
AK075911.1 14 1622 BAC36049.1
AK080777.1 14 1594 BAC38018.1
AK144168.1 14 1749 BAE25744.1
AK150444.1 14 1592 BAE29566.1
AK151960.1 14 1596 BAE30831.1
AK159091.1 15 1592 BAE34808.1
AK159814.1 14 1607 BAE35394.1
AK159890.1 14 1594 BAE35457.1
AK160112.1 14 1581 BAE35636.1
AK167013.1 14 1607 BAE39189.1
AK168186.1 14 1621 BAE40146.1
AK170903.1 14 1608
AK197174.1 2 73
BC010755.1 14 1590 AAH10755.1
U07631.1 14 1613 AAA18775.1
X64331.1 14 1564 CAA45615.1

View Full Product Details