Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mapk15
Entrez Gene ID
332110
Gene Name
mitogen-activated protein kinase 15
Gene Aliases
BC048082
Chromosome Location
Chr.15: 75993731 - 75999153 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177922.2 NP_808590.1 6 - 7 676 66
GenBank mRNA BC048082.1 - 6 - 7 676 66

View Full Product Details