The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008323.1 | NP_032349.1 | 4 - 5 | 277 | 80 |
GenBank mRNA | AJ748655.1 | - | 4 - 5 | 229 | 80 |
AK156309.1 | - | 4 - 5 | 275 | 80 | |
AK166925.1 | - | 3 - 4 | 234 | 80 | |
AK167131.1 | - | 2 - 3 | 805 | 80 | |
AK168526.1 | - | 4 - 5 | 275 | 80 | |
BC085179.1 | - | 4 - 5 | 250 | 80 | |
BC119170.1 | - | 4 - 5 | 260 | 80 | |
BC119172.1 | - | 4 - 5 | 260 | 80 | |
U68564.1 | - | 4 - 5 | 277 | 80 |